Merck
  • Lamiophlomis rotata, an orally available Tibetan herbal painkiller, specifically reduces pain hypersensitivity states through the activation of spinal glucagon-like peptide-1 receptors.
Lamiophlomis rotata, an orally available Tibetan herbal painkiller, specifically reduces pain hypersensitivity states through the activation of spinal glucagon-like peptide-1 receptors.
Anesthesiology (2014-09-24)
Bin Zhu, Nian Gong, Hui Fan, Chong-Sheng Peng, Xiu-Juan Ding, Yi Jiang, Yong-Xiang Wang
ABSTRACT
Lamiophlomis rotata is an orally available Tibetan herb prescribed for the management of pain, with shanzhiside methylester (SM) and 8-O-acetyl-SM as quality control ingredients. This study aimed to evaluate the antinociceptive properties of L. rotata, determine whether SM and 8-O-acetyl-SM are principle effective ingredients, and explore whether L. rotata produces antinociception through activation of spinal glucagon-like peptide-1 receptors (GLP-1Rs). Formalin test, neuropathic pain, and bone cancer pain models were used, and the animal sample size was 5 to 6 in each group. Hydrogen peroxide-induced oxidative damage was also assayed. The L. rotata aqueous extract blocked formalin-induced tonic hyperalgesia and peripheral nerve injury- and bone cancer-induced mechanical allodynia by 50 to 80%, with half-effective doses of 130 to 250 mg/kg, close to the human dosage. The herb was not effective in alleviating acute nociceptive pain. A 7-day gavage with L. rotata aqueous extract did not lead to antiallodynic tolerance. Total iridoid glycosides, rather than total flavonoids, were identified by the activity-tracking method as effective ingredients for antihyperalgesia, whereas both SM and 8-O-acetyl-SM were principal components. Further demonstrations using the GLP-1R antagonist and gene silencer against GLP-1R at both the spinal and the cellular levels indicated that L. rotata inhibited pain hyperactivity by activation of spinal GLP-1Rs, and SM and 8-O-acetyl-SM appeared to be orthosteric, reversible, and fully intrinsic agonists of both rat and human GLP-1Rs. Results support the notion that the activation of spinal GLP-1Rs leads to specific antinociception in pain hypersensitivity and further suggest that GLP-1R is a human-validated target molecule for the treatment of chronic pain.
Product Description
Supelco
Hydrogen peroxide solution, for ultratrace analysis, 30 % (w/w)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Formaldehyde solution, 36.5-38% in H2O