Merck
  • Heteroglucan-dendrimer glycoconjugate: a modulated construct with augmented immune responses and signaling phenomena.
Heteroglucan-dendrimer glycoconjugate: a modulated construct with augmented immune responses and signaling phenomena.
Biochimica et biophysica acta (2014-05-09)
K Sanjana P Devi, Birendra Behera, Banalata Sahoo, Tapas K Maiti
ABSTRACT
Newer strategies for augmenting immune responses of pharmacologically active glucans may serve to improve the medicinal potential of these biomolecules. With this aim, the present work was focused on generating targeted high molecular size glucan particles with magnified immune response activity. Heteroglucans were conjugated with PAMAM dendrimers using a Schiff base reductive amination reaction to generate a polytethered molecule with multiple glucan motifs. The modulated construct was characterized by FTIR, TEM, (1)H NMR and dynamic light scattering (DLS) methods. Effects of conjugated glucans were examined in RAW 264.7 macrophage cells as well as in S-180 murine tumor models. Dendrimer-conjugated glucans were found to exhibit a two-fold increase in immune stimulation in comparison to unconjugated glucans. This may be corroborated by the predominant enhancement in immunological functions such as nitric oxide production, ROS generation and immune directed tumor inhibition in murine models. Immune cell surface markers (CD4, CD8, CD19, MHC-II) and cytokine levels were also found to be highly up-regulated in the splenocytes of mice subjected to particulate glucan administration. Our study also demonstrated that conjugated glucan treatment to RAW 264.7 cells strongly enhanced the phosphorylation of two downstream signalling molecules of the mitogen activated protein kinase (MAPKs) family: p38 and MEK1/2 relative to single glucans thereby relating molecular mechanisms with enhanced immune stimulation. The results obtained thus support that particulate format of soluble heteroglucan will thereby improve its functionality and identify leads in therapeutic competence.
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Monoclonal Anti-SYP antibody produced in mouse, clone 3B3, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Sodium cyanoborohydride solution, 1.0 M in THF
Sigma-Aldrich
Sodium cyanoborohydride, reagent grade, 95%
Sigma-Aldrich
Sodium cyanoborohydride solution, 5.0 M in 1 M NaOH