Merck
  • Induction of autophagy by Tongxinluo through the MEK/ERK pathway protects human cardiac microvascular endothelial cells from hypoxia/reoxygenation injury.
Induction of autophagy by Tongxinluo through the MEK/ERK pathway protects human cardiac microvascular endothelial cells from hypoxia/reoxygenation injury.
Journal of cardiovascular pharmacology (2014-04-08)
Hehe Cui, Xiangdong Li, Na Li, Kang Qi, Qing Li, Chen Jin, Qian Zhang, Leipei Jiang, Yuejin Yang
ABSTRACT
: In contrast to cardiomyocytes, autophagy in cardiac microvascular endothelial cells (CMECs) during ischemia/reperfusion (I/R) injury has not been fully investigated. Tongxinluo (TXL), a traditional Chinese medicine, was shown to be vascular protective. We aimed to elucidate the role of autophagy and its regulatory mechanisms by TXL in CMECs subjected to I/R injury. CMECs were exposed to different treatments for 30 minutes and subjected to hypoxia/reoxygenation each for 2 hours. The results indicated that hypoxia/reoxygenation significantly induced autophagy, as identified by an increased number of monodansylcadaverine-positive CMECs, increased autophagosome formation, and a higher type II/type I of light chain 3 ratio, but not Beclin-1 expression. Autophagy inhibition using 3-methyladenine was proapoptotic, but rapamycin-induced autophagy was antiapoptotic. TXL enhanced autophagy and decreased apoptosis in a dose-dependent manner, reaching its largest effect at 800 μg/mL. 3-methyladenine attenuated the TXL-promoted autophagy and antiapoptotic effects, whereas rapamycin had no additional effects compared with TXL alone. TXL upregulated mitogen-activated protein kinase and extracellular signal-regulated kinase (ERK) phosphorylation; however, PD98059 abrogated ERK phosphorylation and decreased autophagy and increased apoptosis compared with TXL alone. These results suggest that autophagy is a protective mechanism in CMECs subjected to I/R injury and that TXL can promote autophagy through activation of the mitogen-activated protein kinase/ERK pathway.
Sigma-Aldrich
Ethanol, absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC), ≥98% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof
Supelco
Rapamycin, VETRANAL®, analytical standard
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, BioUltra, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.