Merck
  • Apoptosis-regulated low-avidity cancer-specific CD8(+) T cells can be rescued to eliminate HER2/neu-expressing tumors by costimulatory agonists in tolerized mice.
Apoptosis-regulated low-avidity cancer-specific CD8(+) T cells can be rescued to eliminate HER2/neu-expressing tumors by costimulatory agonists in tolerized mice.
Cancer immunology research (2014-04-26)
Chelsea M Black, Todd D Armstrong, Elizabeth M Jaffee
ABSTRACT
A major barrier to vaccines in cancer treatment is their failure to activate and maintain a complete cancer-specific CD8(+) effector T-cell repertoire. Low-avidity T cells are more likely to escape clonal deletion in the thymus when compared with high-avidity T cells, and therefore comprise the major population of effector T cells available for activation in patients with cancer. However, low-avidity T cells fail to traffic into the tumor microenvironment and function in eradicating tumor under optimal vaccination conditions as opposed to high-avidity T cells that escape clonal deletion and function in tumor killing. We used high- and low-avidity T-cell receptor transgenic CD8(+) T cells specific for the immunodominant epitope HER2/neu (RNEU420-429) to identify signaling pathways responsible for the inferior activity of the low-avidity T cells. Adoptive transfer of these cells into tumor-bearing vaccinated mice identified the members of apoptosis pathways that are upregulated in low-avidity T cells. The increased expression of proapoptotic proteins by low-avidity T cells promoted their own cell death and also that of other tumor-specific CD8(+) T cells within their local environment. Importantly, we show that this proapoptotic effect can be overcome by using a strong costimulatory signal that prevents the activation-induced cell death and enables the low-avidity T cells to traffic into the tumor and assist in tumor clearance. These findings identify new therapeutic opportunities for activating the most potent anticancer T-cell responses.
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Brefeldin A, BioXtra, ≥99% (HPLC and TLC)
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO