Merck
  • Cancer stem-like cells derived from chemoresistant tumors have a unique capacity to prime tumorigenic myeloid cells.
Cancer stem-like cells derived from chemoresistant tumors have a unique capacity to prime tumorigenic myeloid cells.
Cancer research (2014-03-19)
Tsunaki Yamashina, Muhammad Baghdadi, Akihiro Yoneda, Ichiro Kinoshita, Shinya Suzu, Hirotoshi Dosaka-Akita, Masahisa Jinushi
ABSTRACT
Resistance to anticancer therapeutics greatly affects the phenotypic and functional properties of tumor cells, but how chemoresistance contributes to the tumorigenic activities of cancer stem-like cells remains unclear. In this study, we found that a characteristic of cancer stem-like cells from chemoresistant tumors (CSC-R) is the ability to produce a variety of proinflammatory cytokines and to generate M2-like immunoregulatory myeloid cells from CD14(+) monocytes. Furthermore, we identified the IFN-regulated transcription factor IRF5 as a CSC-R-specific factor critical for promoting M-CSF production and generating tumorigenic myeloid cells. Importantly, myeloid cells primed with IRF5(+) CSC-R facilitate the tumorigenic and stem cell activities of bulk tumors. Importantly, the activation of IRF5/M-CSF pathways in tumor cells were correlated with the number of tumor-associated CSF1 receptor(+) M2 macrophages in patients with non-small lung cancer. Collectively, our findings show how chemoresistance affects the properties of CSCs in their niche microenvironments.
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)