Merck
  • Obesity-linked homologues TfAP-2 and Twz establish meal frequency in Drosophila melanogaster.
Obesity-linked homologues TfAP-2 and Twz establish meal frequency in Drosophila melanogaster.
PLoS genetics (2014-09-05)
Michael J Williams, Philip Goergen, Jayasimman Rajendran, Galina Zheleznyakova, Maria G Hägglund, Emelie Perland, Sonchita Bagchi, Argyro Kalogeropoulou, Zaid Khan, Robert Fredriksson, Helgi B Schiöth
ABSTRACT
In all animals managing the size of individual meals and frequency of feeding is crucial for metabolic homeostasis. In the current study we demonstrate that the noradrenalin analogue octopamine and the cholecystokinin (CCK) homologue Drosulfakinin (Dsk) function downstream of TfAP-2 and Tiwaz (Twz) to control the number of meals in adult flies. Loss of TfAP-2 or Twz in octopaminergic neurons increased the size of individual meals, while overexpression of TfAP-2 significantly decreased meal size and increased feeding frequency. Of note, our study reveals that TfAP-2 and Twz regulate octopamine signaling to initiate feeding; then octopamine, in a negative feedback loop, induces expression of Dsk to inhibit consummatory behavior. Intriguingly, we found that the mouse TfAP-2 and Twz homologues, AP-2β and Kctd15, co-localize in areas of the brain known to regulate feeding behavior and reward, and a proximity ligation assay (PLA) demonstrated that AP-2β and Kctd15 interact directly in a mouse hypothalamus-derived cell line. Finally, we show that in this mouse hypothalamic cell line AP-2β and Kctd15 directly interact with Ube2i, a mouse sumoylation enzyme, and that AP-2β may itself be sumoylated. Our study reveals how two obesity-linked homologues regulate metabolic homeostasis by modulating consummatory behavior.
Product Description
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%