Merck
  • Immunosuppressive monocytes: possible homeostatic mechanism to restrain chronic intestinal inflammation.
Immunosuppressive monocytes: possible homeostatic mechanism to restrain chronic intestinal inflammation.
Journal of leukocyte biology (2014-04-04)
Elvira Kurmaeva, Dhruva Bhattacharya, Wendy Goodman, Sara Omenetti, Amber Merendino, Seth Berney, Theresa Pizarro, Dmitry V Ostanin
ABSTRACT
Chronic colitis is accompanied by extensive myelopoiesis and accumulation of CD11b+Gr-1+ cells in spleens and secondary lymphoid tissues. Although cells with similar phenotype have been described in cancer, chronic infection, or autoimmunity, where they were associated with suppression of T cell responses, little is known regarding how these cells affect CD4 T cell responses in the context of chronic intestinal inflammation. Therefore, we undertook this study to characterize the interplay between colitis-induced myeloid cells and CD4 T cell. Within the CD11b+Gr-1+ population, only monocytes (Ly6G(neg)Ly6C(high)) but not other myeloid cell subsets suppressed proliferation and production of cytokines by CD4 T cells. Suppression was mediated by cell-contact, NO and partially by IFN-γ and PGs. Interestingly, Ly6C(high) MDCs, isolated from colitic colons, showed up-regulation of iNOS and arginase-1 and were more potent suppressors than those isolated from spleen. On a single-cell level, MDCs inhibited Th1 responses but enhanced generation of foxp3+ T cells. MDCs, cocultured with activated/Teffs, isolated from inflamed colons under hypoxic (1% O2) conditions typical for the inflamed intestine, suppressed proliferation but not their production of proinflammatory cytokines and chemokines. Taken together, expansion of monocytes and MDCs and activation of their suppressive properties may represent a homeostatic mechanism aimed at restraining excessive T cell activation during chronic inflammatory settings. The contribution of immunosuppressive monocytes/MDCs to chronic colitis and their role in shaping T cell responses in vivo require further investigation.
Product Description
Indomethacin, European Pharmacopoeia (EP) Reference Standard
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
L-Arginine
Sigma-Aldrich
NG-Methyl-L-arginine acetate salt, ≥98% (TLC)
Sigma-Aldrich
L-Arginine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Arginine, 99%, FCC, FG
Supelco
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%