Merck
  • Pharmacokinetics and metabolite profiling of fimasartan, a novel antihypertensive agent, in rats.
Pharmacokinetics and metabolite profiling of fimasartan, a novel antihypertensive agent, in rats.
Xenobiotica; the fate of foreign compounds in biological systems (2014-05-03)
Tae Hwan Kim, Soyoung Shin, Mohammad Bashir, Yong Ha Chi, Soo Heui Paik, Joo Han Lee, Hyuk Joon Choi, Jin Ho Choi, Sun Dong Yoo, Jürgen B Bulitta, Eunsook Ma, Sang Hoon Joo, Beom Soo Shin
ABSTRACT
1. The objectives of this study were to evaluate the pharmacokinetics and metabolism of fimasartan in rats. 2. Unlabeled fimasartan or radiolabeled [(14)C]fimasartan was dosed by intravenous injection or oral administration to rats. Concentrations of unlabeled fimasartan in the biological samples were determined by a validated LC/MS/MS assay. Total radioactivity was quantified by liquid scintillation counting and the radioactivity associated with the metabolites was analyzed by using the radiochemical detector. Metabolite identification was conducted by product ion scanning using LC/MS/MS. 3. After oral administration of [(14)C]fimasartan, total radioactivity was found primarily in feces. In bile duct cannulated rats, 58.8 ± 14.4% of the radioactive dose was excreted via bile after oral dosing. Major metabolites of fimasartan including the active metabolite, desulfo-fimasartan, were identified, yet none represented more than 7.2% of the exposure of the parent drug. Fimasartan was rapidly and extensively absorbed and had an oral bioavailability of 32.7-49.6% in rats. Fimasartan plasma concentrations showed a multi-exponential decline after oral administration. Double peaks and extended terminal half-life were observed, which was likely caused by enterohepatic recirculation. 4. These results provide better understanding on the pharmacokinetics of fimasartan and may aid further development of fimasartan analogs.
Sigma-Aldrich
Methanol
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Fimasartan, ≥98% (HPLC)
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Methanol solution, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.