Merck
  • M2 kupffer cells promote hepatocyte senescence: an IL-6-dependent protective mechanism against alcoholic liver disease.
M2 kupffer cells promote hepatocyte senescence: an IL-6-dependent protective mechanism against alcoholic liver disease.
The American journal of pathology (2014-04-10)
Jinghong Wan, Merieme Benkdane, Elizabeth Alons, Sophie Lotersztajn, Catherine Pavoine
ABSTRACT
Alcoholic liver disease is a predominant cause of liver-related mortality in Western countries. The early steps of alcohol-induced steatosis and liver injury involve several mechanisms, including inflammation and oxidative stress. The inflammatory process is initiated by polarization of Kupffer cells toward a proinflammatory M1 phenotype, and we recently found that promoting anti-inflammatory M2 Kupffer cell polarization protects against alcohol-induced hepatocyte steatosis and apoptosis. Alcohol-induced oxidative stress is a potential trigger of senescence, and senescent cells exhibit characteristic functional resistance to apoptosis. We sought to evaluate induction of hepatocyte senescence as an early protective mechanism against alcoholic liver disease. Combining in vivo and in vitro studies, we show that M2 macrophages trigger hepatocyte senescence and enhance alcohol-induced hepatocyte senescence, as indicated by increased β-galactosidase activity, elevated CDKN1A mRNA expression, and induction of nuclear p21. We identify IL-6 as the mediator of M2-induced hepatocyte senescence. Senescent hepatocytes display characteristic resistance to apoptosis but also to steatosis, thus arguing for an early protective effect against alcoholic liver disease. These findings further suggest that pharmacologic interventions targeting M2 polarization during the early stages of alcoholic liver disease may represent an attractive strategy for the limitation of inflammation, hepatocyte apoptosis, and steatosis.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethanol, absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC), ≥98% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof
Sigma-Aldrich
Ethanol, BioUltra, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.