Merck
  • Retinoic acid and hydroquinone induce inverse expression patterns on cornified envelope-associated proteins: implication in skin irritation.
Retinoic acid and hydroquinone induce inverse expression patterns on cornified envelope-associated proteins: implication in skin irritation.
Journal of dermatological science (2014-09-23)
Kyung Ah Cheong, Ha Jung Kim, Ji-Young Kim, Chang-Hyun Kim, Won-Suk Lim, Minsoo Noh, Ai-Young Lee
ABSTRACT
Hydroquinone (HQ) with or without retinoic acid (RA) is routinely used for the treatment of hyperpigmented conditions. Skin irritation is a major problem with popular depigmenting agents, resulting in postinflammatory hyperpigmentation. To examine the molecular mechanism associated with skin irritation by RA or HQ. A genome-wide transcriptional profiling analysis was performed using monolayer cultures of human keratinocytes treated with or without irritant doses of RA, HQ, or sodium lauryl sulfate (SLS), a representative irritant. Differentially expressed genes (DEGs) were mapped on human chromosomes using a Manhattan plot. For the validation of candidate DEGs, the chemicals with different concentrations of varying irritation intensities were applied in vitro and in vivo and analyzed using real time-PCR and Western blotting. DEGs mapped to the 1q21 locus, which is composed of a cluster of genes encoding the cornified envelope precursors, showed an inverse expression pattern in response to HQ and RA. Concentrations of RA and HQ that induced a broad range of irritant responses in cultured cells or mice skin also induced inverse effects on the expression of cornified envelope-associated proteins. Genetic modulation on cornified envelope-associated proteins by RA-induced irritation, which may be involved in physiological skin barrier disturbance, could be inverse to that by HQ- or SLS-induced irritation.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O