Merck
  • Human NK cells licensed by killer Ig receptor genes have an altered cytokine program that modifies CD4+ T cell function.
Human NK cells licensed by killer Ig receptor genes have an altered cytokine program that modifies CD4+ T cell function.
Journal of immunology (Baltimore, Md. : 1950) (2014-06-18)
Lin Lin, Chao Ma, Bo Wei, Najib Aziz, Raja Rajalingam, Susy Yusung, Henry A Erlich, Elizabeth A Trachtenberg, Stephan R Targan, Dermot P B McGovern, James R Heath, Jonathan Braun
ABSTRACT
NK cells are innate immune cells known for their cytolytic activities toward tumors and infections. They are capable of expressing diverse killer Ig-like receptors (KIRs), and KIRs are implicated in susceptibility to Crohn's disease (CD), a chronic intestinal inflammatory disease. However, the cellular mechanism of this genetic contribution is unknown. In this study, we show that the "licensing" of NK cells, determined by the presence of KIR2DL3 and homozygous HLA-C1 in host genome, results in their cytokine reprogramming, which permits them to promote CD4(+) T cell activation and Th17 differentiation ex vivo. Microfluidic analysis of thousands of NK single cells and bulk secretions established that licensed NK cells are more polarized to proinflammatory cytokine production than unlicensed NK cells, including production of IFN-γ, TNF-α, CCL-5, and MIP-1β. Cytokines produced by licensed NK augmented CD4(+) T cell proliferation and IL-17A/IL-22 production. Ab blocking indicated a primary role for IFN-γ, TNF-α, and IL-6 in the augmented T cell-proliferative response. In conclusion, NK licensing mediated by KIR2DL2/3 and HLA-C1 elicits a novel NK cytokine program that activates and induces proinflammatory CD4(+) T cells, thereby providing a potential biologic mechanism for KIR-associated susceptibility to CD and other chronic inflammatory diseases.
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, ≥95.0% (HPLC)