Merck
  • An increased arginase activity is associated with corpus cavernosum impairment induced by hypercholesterolemia.
An increased arginase activity is associated with corpus cavernosum impairment induced by hypercholesterolemia.
The journal of sexual medicine (2014-03-13)
Rodrigo A Fraga-Silva, Fabiana P Costa-Fraga, Younoss Faye, Mikaël Sturny, Robson A S Santos, Rafaela F da Silva, Nikolaos Stergiopulos
ABSTRACT
Hypercholesterolemia is a prevalent risk factor for the development of erectile dysfunction (ED), mostly due to an increase in oxidative stress and impaired nitric oxide (NO) bioavailability within the penis. Arginase is an enzyme that shares the common substrate L-arginine with NO synthase. Augmented arginase activity reduces NO production and is associated with ED development. However, the contribution of arginase hyperactivity in hypercholesterolemia-induced ED is unknown. In the present study, we investigated the activity and role of arginase in the corpus cavernosum of hypercholesterolemic mice. Apolipoprotein E (ApoE) gene-deleted mice fed with a Western-type diet for 11 weeks were treated with the selective arginase inhibitor, N-ω-Hydroxy-L-norarginine (NOHA), or vehicle (saline 0.9%) during the last 9 weeks. Arginase activity and expression were measured in penis protein extraction. Reactive oxygen species (ROS) content within the corpus cavernosum was measured by dihydroethidium staining. Functional in vitro studies were performed using cavernosal strips mounted in an isometric organ bath to evaluate NO production. Arginase activity and its role in modulating NO and ROS production within the corpus cavernosum of hypercholesterolemic mice is the main outcome measure. Total arginase activity and arginase type II protein expression were increased in hypercholesterolemic mice compared with wild-type mice. The long-term treatment with NOHA normalized this alteration. Moreover, pharmacological arginase inhibition by NOHA attenuated the augmented ROS production within the corpus cavernosum of ApoE(-/-) mice, which increased the NO-dependent response in cavernosal strips. These evidences indicate that arginase hyperactivity is associated with ED induced by hypercholesterolemia, suggesting that this enzyme is a potential target for treating ED.
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture