Merck
  • Identification of UACA, EXOSC9, and ΤΜX2 in bovine periosteal cells by mass spectrometry and immunohistochemistry.
Identification of UACA, EXOSC9, and ΤΜX2 in bovine periosteal cells by mass spectrometry and immunohistochemistry.
Analytical and bioanalytical chemistry (2014-04-04)
Mari Akiyama
ABSTRACT
Inspection of patient-derived cells used in transplantation is non-invasive. Therefore, proteomics analysis using supernatants of cells cultured before transplantation is informative. In order to investigate the cell niche of bovine periosteal cells, supernatants of these cultured cells were subjected to 2-D electrophoresis followed by mass spectrometry, which identified type 1 collagen and the C-terminus of type 3 collagen. Only the C-terminal peptide from type 3 collagen was found in supernatants. It is known that type 3 collagen may be expressed intra- or extra-cellularly. Paraffin sections of the cultured cells were next examined by immunohistochemistry, which revealed that type 3 collagen regions besides the C-terminal peptide were present around the bovine periosteal cells but were not found in supernatants. Full-length type 3 collagen was closely associated with the cells, and only the C-terminal peptide was detectable in culture supernatants. Mass spectrometry analysis of partial peptide data combined with immunohistochemistry also indicated that uveal autoantigen with coiled coil domains and ankyrin repeats (UACA), exosome complex component RRP45 (EXOSC9), and thioredoxin-related transmembrane protein 2 (TMX2) were expressed in bovine periosteal cells. Results of this study indicate that analysis of culture supernatants before cell transplantation can provide useful biomarkers indicating the niche of cells used for transplantation.
Product Description
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Urea, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, powder, BioReagent, suitable for cell culture
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, BioUltra, 99% (T)
Supelco
Urea, analytical standard
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Glycine, European Pharmacopoeia (EP) Reference Standard
SAFC
Glycine
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O