Merck
  • The effect of temperature on the catalytic conversion of Kraft lignin using near-critical water.
The effect of temperature on the catalytic conversion of Kraft lignin using near-critical water.
Bioresource technology (2014-08-20)
Thi Dieu Huyen Nguyen, Marco Maschietti, Lars-Erik Åmand, Lennart Vamling, Lars Olausson, Sven-Ingvar Andersson, Hans Theliander
ABSTRACT
The catalytic conversion of suspended LignoBoost Kraft lignin was performed in near-critical water using ZrO2/K2CO3 as the catalytic system and phenol as the co-solvent and char suppressing agent. The reaction temperature was varied from 290 to 370°C and its effect on the process was investigated in a continuous flow (1kg/h). The yields of water-soluble organics (WSO), bio-oil and char (dry lignin basis) were in the ranges of 5-11%, 69-87% and 16-22%, respectively. The bio-oil, being partially deoxygenated, exhibited higher carbon content and heat value, but lower sulphur content than lignin. The main 1-ring aromatics (in WSO and diethylether-soluble bio-oil) were anisoles, alkylphenols, catechols and guaiacols. The results show that increasing temperature increases the yield of 1-ring aromatics remarkably, while it increases the formation of char moderately. An increase in the yields of anisoles, alkylphenols and catechols, together with a decrease in the yield of guaiacols, was also observed.
Product Description
Supelco
4-Hydroxybenzaldehyde, analytical standard
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
4-Hydroxybenzaldehyde, ≥95.0% (HPLC)
Supelco
o-Cresol, analytical standard
Sigma-Aldrich
o-Cresol, ≥99%
Sigma-Aldrich
4-Hydroxybenzaldehyde, 98%
Sigma-Aldrich
p-Cresol, ≥99%, FG
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Sigma-Aldrich
4-Hydroxybenzaldehyde, ≥97%, FG
Supelco
Guaiacol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Guaiacol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Guaiacol, oxidation indicator
Sigma-Aldrich
p-Cresol, 99%
Supelco
p-Cresol, analytical standard
Sigma-Aldrich
o-Cresol, ReagentPlus®, ≥99%
Supelco
Anisole, analytical standard
Sigma-Aldrich
Anisole, ReagentPlus®, 99%
Supelco
Anisole, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
m-Cresol, ≥98%, FG
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Anisole, ≥99%, FCC, FG
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Supelco
m-Cresol, analytical standard