Merck
  • Control of food intake and energy expenditure by Nos1 neurons of the paraventricular hypothalamus.
Control of food intake and energy expenditure by Nos1 neurons of the paraventricular hypothalamus.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-11-14)
Amy K Sutton, Hongjuan Pei, Korri H Burnett, Martin G Myers, Christopher J Rhodes, David P Olson
ABSTRACT
The paraventricular nucleus of the hypothalamus (PVH) contains a heterogeneous cluster of Sim1-expressing cell types that comprise a major autonomic output nucleus and play critical roles in the control of food intake and energy homeostasis. The roles of specific PVH neuronal subtypes in energy balance have yet to be defined, however. The PVH contains nitric oxide synthase-1 (Nos1)-expressing (Nos1(PVH)) neurons of unknown function; these represent a subset of the larger population of Sim1-expressing PVH (Sim1(PVH)) neurons. To determine the role of Nos1(PVH) neurons in energy balance, we used Cre-dependent viral vectors to both map their efferent projections and test their functional output in mice. Here we show that Nos1(PVH) neurons project to hindbrain and spinal cord regions important for food intake and energy expenditure control. Moreover, pharmacogenetic activation of Nos1(PVH) neurons suppresses feeding to a similar extent as Sim1(PVH) neurons, and increases energy expenditure and activity. Furthermore, we found that oxytocin-expressing PVH neurons (OXT(PVH)) are a subset of Nos1(PVH) neurons. OXT(PVH) cells project to preganglionic, sympathetic neurons in the thoracic spinal cord and increase energy expenditure upon activation, though not to the same extent as Nos1(PVH) neurons; their activation fails to alter feeding, however. Thus, Nos1(PVH) neurons promote negative energy balance through changes in feeding and energy expenditure, whereas OXT(PVH) neurons regulate energy expenditure alone, suggesting a crucial role for non-OXT Nos1(PVH) neurons in feeding regulation.
Product Description
Supelco
Hydrogen peroxide solution, for ultratrace analysis, 30 % (w/w)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat
Sigma-Aldrich
(2-Hydroxypropyl)-β-cyclodextrin, powder, BioReagent, suitable for cell culture