Merck
  • Comparative analysis of Hsp10 and Hsp90 expression in healthy mucosa and adenocarcinoma of the large bowel.
Comparative analysis of Hsp10 and Hsp90 expression in healthy mucosa and adenocarcinoma of the large bowel.
Anticancer research (2014-07-31)
Francesca Rappa, Carmelo Sciume, Margherita Lo Bello, Celeste Caruso Bavisotto, Antonella Marino Gammazza, Rosario Barone, Claudia Campanella, Sabrina David, Francesco Carini, Federica Zarcone, Stefano Rizzuto, Adriana Lena, Giovanni Tomasello, Maria Laura Uzzo, Giovanni Francesco Spatola, Giuseppe Bonaventura, Angelo Leone, Aldo Gerbino, Francesco Cappello, Fabio Bucchieri, Giovanni Zummo, Felicia Farina
ABSTRACT
Heat shock proteins (Hsps) assist other proteins in their folding and drive the degradation of defective proteins. During evolution, these proteins have also acquired other roles. Hsp10 is involved in immunomodulation and tumor progression. Hsp90 stabilizes a range of "client" proteins involved in cell signaling. The present study evaluated the expression levels of Hsp10 and Hsp90 in normal mucosa and adenocarcinoma samples of human large bowel. Samples of normal mucosa and adenocarcinoma were collected and Reverse transcriptase-polymerase chain reaction RT-PCR, western blotting (WB) analyses, as well as immunohistochemistry were performed to evaluate the expression levels of Hsp10 and Hsp90. RT-PCR showed a higher gene expression of Hsp10 and Hsp90 in adenocarcinoma samples compared to healthy mucosa. WB results confirmed these findings. Immunohistochemistry revealed higher levels of Hsp10 in adenocarcinoma in both the epithelium and the lamina propria, while Hsp90 expression was higher in the adenocarcinoma samples only in the lamina propria. Hsp10 and Hsp90 may be involved in large bowel carcinogenesis.
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
TRI Reagent®, For processing tissues, cells cultured in monolayer or cell pellets
Sigma-Aldrich
Hematoxylin