Merck
  • Sequential Cdk1 and Plk1 phosphorylation of caspase-8 triggers apoptotic cell death during mitosis.
Sequential Cdk1 and Plk1 phosphorylation of caspase-8 triggers apoptotic cell death during mitosis.
Molecular oncology (2014-02-04)
Yves Matthess, Monika Raab, Rainald Knecht, Sven Becker, Klaus Strebhardt
ABSTRACT
Caspase-8 is crucial for cell death induction, especially via the death receptor pathway. The dysregulated expression or function of caspase-8 can promote tumor formation, progression and treatment resistance in different human cancers. Here, we show procaspase-8 is regulated during the cell cycle through the concerted inhibitory action of Cdk1/cyclin B1 and polo-like kinase 1 (Plk1). By phosphorylating S387 in procaspase-8 Cdk1/cyclin B1 generates a phospho-epitope for the binding of the PBD of Plk1. Subsequently, S305 in procaspase-8 is phosphorylated by Plk1 during mitosis. Using an RNAi-based strategy we could demonstrate that the extrinsic cell death is increased upon Fas-stimulation when endogenous caspase-8 is replaced by a mutant (S305A) mimicking the non-phosphorylated form. Together, our data show that sequential phosphorylation by Cdk1/cyclin B1 and Plk1 decreases the sensitivity of cells toward stimuli of the extrinsic pathway during mitosis. Thus, the clinical Plk1 inhibitor BI 2536 decreases the threshold of different cancer cell types toward Fas-induced cell death.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, clone 3H10, clone 3H10, Upstate®, from mouse
Sigma-Aldrich
Anti-PLK1 Antibody, clone 35-206, clone 35-206, Upstate®, from mouse
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone VIN-11-5, ascites fluid
Sigma-Aldrich
CelLytic B Cell Lysis Reagent, For bacterial cell lysis, standard strength