Merck
  • The aldosterone receptor antagonist spironolactone prevents peritoneal inflammation and fibrosis.
The aldosterone receptor antagonist spironolactone prevents peritoneal inflammation and fibrosis.
Laboratory investigation; a journal of technical methods and pathology (2014-05-28)
Lei Zhang, Jian-Bing Hao, Lian-Sheng Ren, Jiu-Li Ding, Li-Rong Hao
ABSTRACT
Peritoneal fibrosis is a complication of patients with long-term continuous ambulatory peritoneal dialysis (CAPD). Reports have indicated that angiotensin (Ang) II may correlate with the development of peritoneal fibrosis. However, it is unknown whether aldosterone also has a role in the development of peritoneal inflammation and fibrosis. The aim of the present study was to clarify the role of aldosterone in peritoneal inflammation and fibrosis. A rat model of peritoneal inflammation and fibrosis was established by daily intraperitoneal injection of dialysates and lipopolysaccharide in a 4-day interval over a period of 7 days. The animals were randomly assigned to five groups as follows: control (C); peritoneal dialysis (PD); peritoneal dialysis-spironolactone (PD-S); peritoneal dialysis-cilazapril (PD-C); and peritoneal dialysis-spironolactone-cilazapril (PD-SC). After 30 days, the TGF-β1 concentration in dialysates from all treatment groups was determined by ELISA. The histopathology of the parietal peritoneum was examined, and the expression of MCP-1, c-Jun, fibronectin (FN) and TGF-β1 in the abdominal membrane was determined by immunohistochemistry. Mineralocorticoid receptor (MR), 11β-hydroxysteroid dehydrogenase type 2 (11β-HSD2) and CYP11B2 (aldosterone synthase) were analyzed by real time-PCR. Collagen deposition was significantly higher in PD compared with the other groups. The expression of MR, 11β-HSD2 and CYP11B2 was significantly higher in PD compared with the other groups. Spironolactone and/or cilazapril treatment partially ablated the increase in monocyte chemoattractant protein (MCP)-1, p-c-Jun, transforming growth factor (TGF)-β1, FN, MR, 11β-HSD2 and CYP11B2. Furthermore, treatment with spironolactone and/or cilazapril also reduced the infiltration of CD-4- and ED-1-positive cells in rat peritoneal tissues after peritoneal fibrosis. Exogenous aldosterone may have a key role in the development of peritoneal inflammation and fibrosis. Spironolactone decreased peritoneal inflammation and fibrosis, which was associated with reduced secretion from peritoneal macrophages, inactivation of the c-Jun N-terminal kinase (JNK) pathway and subsequent downregulation of the expression of TGF-β1.
Sigma-Aldrich
Aldosterone, ≥95% (HPLC)
Cilazapril, European Pharmacopoeia (EP) Reference Standard
Spironolactone for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Aldosterone solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Cilazapril impurity D, European Pharmacopoeia (EP) Reference Standard
Spironolactone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Spironolactone, 97.0-103.0%