Merck
  • Reduced cellular Mg²⁺ content enhances hexose 6-phosphate dehydrogenase activity and expression in HepG2 and HL-60 cells.
Reduced cellular Mg²⁺ content enhances hexose 6-phosphate dehydrogenase activity and expression in HepG2 and HL-60 cells.
Archives of biochemistry and biophysics (2014-03-19)
Chesinta Voma, Andrew Barfell, Colleen Croniger, Andrea Romani
ABSTRACT
We have reported that Mg(2+) dynamically regulates glucose 6-phosphate entry into the endoplasmic reticulum and its hydrolysis by the glucose 6-phosphatase in liver cells. In the present study, we report that by modulating glucose 6-phosphate entry into the endoplasmic reticulum of HepG2 cells, Mg(2+) also regulates the oxidation of this substrate via hexose 6-phosphate dehydrogenase (H6PD). This regulatory effect is dynamic as glucose 6-phosphate entry and oxidation can be rapidly down-regulated by the addition of exogenous Mg(2+). In addition, HepG2 cells growing in low Mg(2+) show a marked increase in hexose 6-phosphate dehydrogenase mRNA and protein expression. Metabolically, these effects on hexose 6-phosphate dehydrogenase are important as this enzyme increases intra-reticular NADPH production, which favors fatty acid and cholesterol synthesis. Similar effects of Mg(2+) were observed in HL-60 cells. These and previously published results suggest that in an hepatocyte culture model changes in cytoplasmic Mg(2+) content regulates glucose 6-phosphate utilization via glucose 6 phosphatase and hexose-6 phosphate dehydrogenase in alternative to glycolysis and glycogen synthesis. This alternative regulation might be of relevance in the transition from fed to fasted state.
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Magnesium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, wire reel, 2m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, wire reel, 1m, diameter 0.125mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, wire reel, 2m, diameter 1.0mm, as drawn, 99.9+%
Magnesium, wire reel, 1m, diameter 0.25mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 1.1mm, as drawn, 99.9+%
Magnesium, microfoil, 25x25mm, thinness 1.0μm, specific density 174μg/cm2, 6 micron aluminum permanent support, 99.9%
Magnesium, wire reel, 0.5m, diameter 0.125mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 1.1mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 0.4mm, as drawn, 99.9+%
Magnesium, microleaf, 25x25mm, thinness 1.0μm, specific density 174μg/cm2, removable support, 99.9%
Magnesium, wire reel, 1m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, wire reel, 2m, diameter 0.4mm, as drawn, 99.9+%
Magnesium, wire reel, 2m, diameter 0.125mm, as drawn, 99.9+%
Sigma-Aldrich
Magnesium, grit, ≥99.0% (KT)
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Sigma-Aldrich
D-Glucose 6-phosphate solution, ~1 M in H2O (approx. 260 mg per ml)
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Sigma-Aldrich
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
Magnesium, 20-230 mesh, reagent grade, 98%
Sigma-Aldrich
Magnesium, powder, ≥99%
Sigma-Aldrich
Magnesium, turnings, 5-25 mm, 99.95% trace metals basis
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol