Merck
  • Fungal morphogenetic pathways are required for the hallmark inflammatory response during Candida albicans vaginitis.
Fungal morphogenetic pathways are required for the hallmark inflammatory response during Candida albicans vaginitis.
Infection and immunity (2014-01-31)
Brian M Peters, Glen E Palmer, Andrea K Nash, Elizabeth A Lilly, Paul L Fidel, Mairi C Noverr
ABSTRACT
Vulvovaginal candidiasis, caused primarily by Candida albicans, presents significant health issues for women of childbearing age. As a polymorphic fungus, the ability of C. albicans to switch between yeast and hyphal morphologies is considered its central virulence attribute. Armed with new criteria for defining vaginitis immunopathology, the purpose of this study was to determine whether the yeast-to-hypha transition is required for the hallmark inflammatory responses previously characterized during murine vaginitis. Kinetic analyses of vaginal infection with C. albicans in C57BL/6 mice demonstrated that fungal burdens remained constant throughout the observation period, while polymorphonuclear leukocyte (PMN), S100A8, and interleukin-1β levels obtained from vaginal lavage fluid increased by day 3 onward. Lactate dehydrogenase activity was also positively correlated with increased effectors of innate immunity. Additionally, immunodepletion of neutrophils in infected mice confirmed a nonprotective role for PMNs during vaginitis. Determination of the importance of fungal morphogenesis during vaginitis was addressed with a two-pronged approach. Intravaginal inoculation of mice with C. albicans strains deleted for key transcriptional regulators (bcr1Δ/Δ, efg1Δ/Δ, cph1Δ/Δ, and efg1Δ/Δ cph1Δ/Δ) controlling the yeast-to-hypha switch revealed a crucial role for morphogenetic signaling through the Efg1 and, to a lesser extent, the Bcr1 pathways in contributing to vaginitis immunopathology. Furthermore, overexpression of transcription factors NRG1 and UME6, to maintain yeast and hyphal morphologies, respectively, confirmed the importance of morphogenesis in generating innate immune responses in vivo. These results highlight the yeast-to-hypha switch and the associated morphogenetic response as important virulence components for the immunopathogenesis of Candida vaginitis, with implications for transition from benign colonization to symptomatic infection.
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type II, ammonium sulfate suspension, 800-1,200 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type XI, lyophilized powder, 600-1,200 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from porcine heart, ammonium sulfate suspension, ≥200 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, Type III, ammonium sulfate suspension, ≥500 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine muscle, Type X, ammonium sulfate suspension, ≥600 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, Type XVII, buffered aqueous glycerol solution, ≥400 units/mg protein
Sigma-Aldrich
Lactic Dehydrogenase, recombinant from E. coli, ≥90 U/mg
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, 1000 units/mL