Merck
  • Expanding the repertoire of plasmids for PCR-mediated epitope tagging in yeast.
Expanding the repertoire of plasmids for PCR-mediated epitope tagging in yeast.
Yeast (Chichester, England) (2008-03-14)
Zarmik Moqtaderi, Kevin Struhl
ABSTRACT
Epitope tagging of yeast proteins provides a convenient means of tracking proteins of interest in Western blots and immunoprecipitation experiments without the need to raise and test specific antibodies. We have constructed four plasmids for use as templates in PCR-based epitope tagging in the yeast Saccharomyces cerevisiae. These plasmids expand the range of epitopes available in a tag-URA3-tag context to include the FLAG, HSV, V5 and VSV-G epitopes. The cloning strategy used would be easily applicable to the construction of a similar tag-URA3-tag molecule for essentially any desired epitope. Oligonucleotides designed for PCR from one plasmid may be used interchangeably with any of the other template molecules to allow tagging with different epitopes without the need for new primer synthesis. We have tagged Tfc6 with each of the triple epitope tags and assessed the efficiency of these epitopes for chromatin immunoprecipitation (ChIP). For all the tagged alleles, ChIP occupancy signals are easily detectable at known Tfc6 target genes. These new tags provide additional options in experimental schemes requiring multiple tagged proteins.
Sigma-Aldrich
Anti-V5 antibody, Mouse monoclonal, clone V5-10, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-HSV antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution