Merck
  • 1,5-Diphenylpent-3-en-1-ynes and methyl naphthalene carboxylates from Lawsonia inermis and their anti-inflammatory activity.
1,5-Diphenylpent-3-en-1-ynes and methyl naphthalene carboxylates from Lawsonia inermis and their anti-inflammatory activity.
Phytochemistry (2013-01-29)
Jing-Ru Liou, Mohamed El-Shazly, Ying-Chi Du, Chao-Neng Tseng, Tsong-Long Hwang, Yueh-Lin Chuang, Yu-Ming Hsu, Pei-Wen Hsieh, Chin-Chun Wu, Shu-Li Chen, Ming-Feng Hou, Fang-Rong Chang, Yang-Chang Wu
ABSTRACT
Lawsonia inermis (Lythraceae) known as henna is one of the most popular and ancient plants used in cosmetics and hair dying. It is cultivated for its leaves but other parts such as seeds, flowers, stem bark and roots are also used in traditional medicine for millennia. Henna tattoo paste also proved to be beneficial for wound healing and in several skin diseases suggesting potent anti-inflammatory activity. To evaluate henna anti-inflammatory activity, 31 compounds, including three 1,5-diphenylpent-3-en-1-yne derivatives, lawsochylin A-C and three methyl naphthalene carboxylates, lawsonaphthoate A-C, were isolated from the stems and leaves of henna utilizing a bioassay-guided fractionation. The structures of the compounds were elucidated by spectroscopic data. Two compounds, lawsochylin A and lawsonaphthoate A showed potent anti-inflammatory activity by inhibition of superoxide anion generation (IC(50)=1.80 and 1.90 μg/ml) and elastase release (IC(50)=1.58 and 3.17 μg/ml) of human neutrophils in response to fMLP or cytochalasin B. Moreover, the known compounds, luteolin, apigenin, 4S-4-hydroxy-α-tetralone, and 2-butoxysuccinic acid, also showed potent inhibition of superoxide anion generation (IC(50)=0.75-1.78 μg/ml) and elastase release (IC(50)=1.62-3.61 μg/ml).
Product Description
Supelco
Naphthalene, analytical standard
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Supelco
Naphthalene solution, certified reference material, 5000 μg/mL in methanol
Supelco
Naphthalene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Naphthalene, 99%
Supelco
Melting point standard 79-81°C, analytical standard