Merck
  • Evaluation of cannabinoid receptor 2 and metabotropic glutamate receptor 1 functional responses using a cell impedance-based technology.
Evaluation of cannabinoid receptor 2 and metabotropic glutamate receptor 1 functional responses using a cell impedance-based technology.
Journal of biomolecular screening (2010-09-03)
Paola Scandroglio, Rossella Brusa, Gianluca Lozza, Isabella Mancini, Roberta Petrò, Angelo Reggiani, Massimiliano Beltramo
ABSTRACT
Recently, new technologies based on biosensors and called label free have been developed. These technologies eliminate the need for using markers and dyes. The authors applied one of these technologies, based on measurement of cell impedance variation, to study the pharmacological profiles of ligands for the cannabinoid receptor 2 (CB2), a Gi-coupled receptor, and for the metabopotropic glutamate receptor 1 (mGluR1), a Gq-coupled receptor. Reference agonists and antagonists/inverse agonists for the 2 receptors were applied to recombinant cell lines and impedance monitored over time. Agonists (JWH133 and CP55940 for CB2; quisqualate, glutamate, 1S-3R-ACPD, and S-3,5-DHPG for mGluR1) triggered a variation of impedance consistent in both potency and efficacy with data obtained using classical assays measuring cAMP or Ca(2+) levels. This effect was not present in the parental nontransfected cell line, confirming specific receptor-mediated response. Application of antagonists (AM630 for CB2; YM298198, SCH1014222, J&J16259685, and CPCCOEt for mGluR1) reduced agonist-induced impedance changes. The only exception was the mGluR1 antagonist BAY367620 that, while active in the Ca(2+) assay, was inactive in the impedance assay. Overall, these results confirm the possibility of using cell impedance-based technology to study the pharmacological profile of ligands acting at G-protein-coupled receptors coupled to different downstream signaling pathways.
Sigma-Aldrich
BAY 36-7620, ≥98% (HPLC)
Sigma-Aldrich
Cycloleucine, 97%
Sigma-Aldrich
Quisqualic acid, powder
Sigma-Aldrich
trans-(±)-ACPD monohydrate, solid, ≥97% (NMR)
Sigma-Aldrich
Probenecid