Merck
  • Effect of cadmium injected in ovo on hatching results and the activity of plasma hydrolytic enzymes in newly hatched chicks.
Effect of cadmium injected in ovo on hatching results and the activity of plasma hydrolytic enzymes in newly hatched chicks.
Acta veterinaria Hungarica (2011-07-06)
Małgorzata Dżugan, Marcin Lis, Maria Droba, Jerzy W Niedziółka
ABSTRACT
The aim of the study was to determine the toxicity of cadmium ions in chick embryos, using plasma hydrolytic enzyme as its biomarker. Hatching eggs (n = 300) from Ross 308 broilers were incubated under standard conditions. On day 4 of incubation, 50 μl of saline solution, containing Cd ions at a concentration from 0 (control group) to 24 μg, was injected in ovo into the egg albumen. The results indicate that the administration of cadmium at doses exceeding 1 μg/egg caused a gradual decrease in hatchability, with an LD50 of 3.9 μg/egg. The greatest differences between the groups in the enzymatic activities studied were found for N-acetyl-β-D-glucosaminidase (NAG), β-D-mannosidase (β-MAN) and arylsulphatase (ARYL). Compared to the control group, in the blood serum of chicks from the groups receiving 3, 6 and 12 μg Cd/egg the NAG activity increased by 79, 108 and 54% and β-MAN activity by 33, 119 and 108%, respectively. Exposure to cadmium at a dose of 1 to 6 μg per egg caused an about 60% increase in ARYL activity while a dose of 12 μg decreased the activity by about 35% below the level observed in the control group. These findings show that cadmium has a similar toxicity mechanism in mammals and birds, which opens the possibility of using NAG activity as a biomarker of the cytotoxic effect of cadmium in birds.
Sigma-Aldrich
Sulfatase from abalone entrails, Type VIII, lyophilized powder, 20-40 units/mg solid
Sigma-Aldrich
Sulfatase from Helix pomatia, Type H-2, aqueous solution, ≥2,000 units/mL
Sigma-Aldrich
Sulfatase from Helix pomatia, Type H-1, sulfatase ≥10,000 units/g solid
Sigma-Aldrich
Sulfatase from Patella vulgata (keyhole limpet), Type IV, essentially salt-free, lyophilized powder, ≥10 units/mg solid
Sigma-Aldrich
Sulfatase from Aerobacter aerogenes, Type VI, buffered aqueous glycerol solution, 2-5 units/mg protein (biuret), 10-20 units/mL