Merck
  • CCR5 signaling promotes lipopolysaccharide-induced macrophage recruitment and alveolar developmental arrest.
CCR5 signaling promotes lipopolysaccharide-induced macrophage recruitment and alveolar developmental arrest.
Cell death & disease (2021-02-17)
Ze Chen, Xiaohua Xie, Na Jiang, Jianhui Li, Lei Shen, Yongjun Zhang
ABSTRACT
The pathogenesis of bronchopulmonary dysplasia (BPD), involves inflammatory, mechanisms that are not fully characterized. Here we report that overexpression of C-C chemokine receptor 5 (CCR5) and its ligands is associated with BPD development. Lipopolysaccharide-induced BPD rats have increased CCR5 and interleukin-1β (IL-1β) levels, and decreased alveolarization, while CCR5 or IL-1β receptor antagonist treatments decreased inflammation and increased alveolarization. CCR5 enhances macrophage migration, macrophage infiltration in the lungs, IL-1β levels, lysyl oxidase activity, and alveolar development arrest. CCR5 expression on monocytes, and its ligands in blood samples from BPD infants, are elevated. Furthermore, batyl alcohol supplementation reduced CCR5 expression and IL-1β production in lipopolysaccharide-exposed rat lungs. Moreover, receptor-interacting kinase 3 (RIP3) upstream regulator of CCR5-cultured RIP3-/- macrophages exhibited partly blocked lipopolysaccharide-induced CCR5 expression. We conclude that increased CCR5 expression is a key mechanism in BPD development and represents a novel therapeutic target for treatment.
Sigma-Aldrich
3,7-Diacetyl-1,3,7-triaza-5-phosphabicyclo[3.3.1]nonane, 97%
Sigma-Aldrich
DAPTA trifluoroacetate, ≥95% (HPLC)
Sigma-Aldrich
4,4,4-Trifluoro-1-phenyl-1,3-butanedione, 99%