Merck
  • Small Molecule SARM1 Inhibitors Recapitulate the SARM1-/- Phenotype and Allow Recovery of a Metastable Pool of Axons Fated to Degenerate.
Small Molecule SARM1 Inhibitors Recapitulate the SARM1-/- Phenotype and Allow Recovery of a Metastable Pool of Axons Fated to Degenerate.
Cell reports (2021-01-07)
Robert O Hughes, Todd Bosanac, Xianrong Mao, Thomas M Engber, Aaron DiAntonio, Jeffrey Milbrandt, Rajesh Devraj, Raul Krauss
ABSTRACT
Axonal degeneration is responsible for disease progression and accumulation of disability in many neurodegenerative conditions. The axonal degenerative process can generate a metastable pool of damaged axons that remain structurally and functionally viable but fated to degenerate in the absence of external intervention. SARM1, an NADase that depletes axonal energy stores upon activation, is the central driver of an evolutionarily conserved program of axonal degeneration. We identify a potent and selective small molecule isoquinoline inhibitor of SARM1 NADase that recapitulates the SARM1-/- phenotype and protects axons from degeneration induced by axotomy or mitochondrial dysfunction. SARM1 inhibition post-mitochondrial injury with rotenone allows recovery and rescues axons that already entered the metastable state. We conclude that SARM1 inhibition with small molecules has the potential to treat axonopathies of the central and peripheral nervous systems by preventing axonal degeneration and by allowing functional recovery of a metastable pool of damaged, but viable, axons.
Sigma-Aldrich
5-Fluoro-2′-deoxyuridine, thymidylate synthase inhibitor
Sigma-Aldrich
Rotenone, A mitochondrial toxin and a potent, reversible, and competitive inhibitor of complex I (NADH-CoQ reductase) of the respiratory chain.
Sigma-Aldrich
2-Chloroadenosine
Sigma-Aldrich
DSRM-3716, ≥98% (HPLC)
Sigma-Aldrich
Anti-APP A4 Antibody, a.a. 66-81 of APP {NT}, clone 22C11, clone 22C11, Chemicon®, from mouse
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid