Merck
  • An optimized LC-MS/MS method as a pivotal tool to steer thiamine biofortification strategies in rice.
An optimized LC-MS/MS method as a pivotal tool to steer thiamine biofortification strategies in rice.
Talanta (2021-01-01)
Jana Verstraete, Simon Strobbe, Dominique Van Der Straeten, Christophe Stove
ABSTRACT
In developing countries, people mainly depend on rice as their primary source of calories. However, the thiamine content of rice is below minimal requirements. Biofortification, via genetic engineering, is a cost-effective strategy to increase thiamine content in rice. We report on the optimization of a matrix-specific method, including extensive optimization of the sample preparation to ensure maximal sensitivity and stability. The LC-MS/MS method was fully validated for the simultaneous quantification of thiamine, its precursors 4-methyl-5-(2-hydroxyethyl) thiazole (HET) and 4-amino-2-methyl-5-hydroxymethylpyrimidine (HMP) and its diphosphate derivative (TDP) in both polished and unpolished rice. Bias was below 9% for all analytes and total imprecision (CV%) was within pre-set acceptance criteria (≤15%) for both QCs and real samples. Thiamine monophosphate (TMP), for which no labeled analogue was available at the time of analysis, was determined without internal standard. Although both accuracy and precision criteria were met (bias and CV < 12%), the determination of TMP was considered semi-quantitatively. Moreover, TMP was found to be only a minor thiamine form (<1% of total thiamine in all lines analyzed, both wild-type and genetically engineered), with measurable levels only present in unpolished rice. Finally, the validity and applicability of the procedure were demonstrated via its successful application on rice lines, genetically engineered to enhance thiamine content. Consequently, this method allows to evaluate the success of biofortification strategies in rice.
Sigma-Aldrich
Phosphatase, Acid from sweet potato, ammonium sulfate suspension, ≥10.0 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Protease from Streptomyces griseus, Type XIV, ≥3.5 units/mg solid, powder
Sigma-Aldrich
Thymidine 5′-monophosphate disodium salt hydrate, ≥99%
Sigma-Aldrich
α-Amylase from porcine pancreas, PMSF Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
Sigma-Aldrich
2-Amino-4-hydroxy-6-methylpyrimidine, 98%