Merck
  • TRIB3 promotes MYC-associated lymphoma development through suppression of UBE3B-mediated MYC degradation.
TRIB3 promotes MYC-associated lymphoma development through suppression of UBE3B-mediated MYC degradation.
Nature communications (2020-12-11)
Ke Li, Feng Wang, Zhao-Na Yang, Ting-Ting Zhang, Yu-Fen Yuan, Chen-Xi Zhao, Zaiwuli Yeerjiang, Bing Cui, Fang Hua, Xiao-Xi Lv, Xiao-Wei Zhang, Jiao-Jiao Yu, Shan-Shan Liu, Jin-Mei Yu, Shuang Shang, Yang Xiao, Zhuo-Wei Hu
ABSTRACT
The transcription factor MYC is deregulated in almost all human cancers, especially in aggressive lymphomas, through chromosomal translocation, amplification, and transcription hyperactivation. Here, we report that high expression of tribbles homologue 3 (TRIB3) positively correlates with elevated MYC expression in lymphoma specimens; TRIB3 deletion attenuates the initiation and progression of MYC-driven lymphoma by reducing MYC expression. Mechanistically, TRIB3 interacts with MYC to suppress E3 ubiquitin ligase UBE3B-mediated MYC ubiquitination and degradation, which enhances MYC transcriptional activity, causing high proliferation and self-renewal of lymphoma cells. Use of a peptide to disturb the TRIB3-MYC interaction together with doxorubicin reduces the tumor burden in MycEμ mice and patient-derived xenografts. The pathophysiological relevance of UBE3B, TRIB3 and MYC is further demonstrated in human lymphoma. Our study highlights a key mechanism for controlling MYC expression and a potential therapeutic option for treating lymphomas with high TRIB3-MYC expression.
Sigma-Aldrich
Anti-β-Catenin Antibody, Upstate®, from rabbit
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Formaldehyde solution, 36.5-38% in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting human TRIB3
Supelco
Tamoxifen, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit
Sigma-Aldrich
N-Acetyl-D-lactosamine, ≥98% (TLC)
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)