Merck
  • PRL3-DDX21 Transcriptional Control of Endolysosomal Genes Restricts Melanocyte Stem Cell Differentiation.
PRL3-DDX21 Transcriptional Control of Endolysosomal Genes Restricts Melanocyte Stem Cell Differentiation.
Developmental cell (2020-07-12)
Jeanette A Johansson, Kerrie L Marie, Yuting Lu, Alessandro Brombin, Cristina Santoriello, Zhiqiang Zeng, Judith Zich, Philippe Gautier, Alex von Kriegsheim, Hannah Brunsdon, Ann P Wheeler, Marcel Dreger, Douglas R Houston, Christopher M Dooley, Andrew H Sims, Elisabeth M Busch-Nentwich, Leonard I Zon, Robert S Illingworth, E Elizabeth Patton
ABSTRACT
Melanocytes, replenished throughout life by melanocyte stem cells (MSCs), play a critical role in pigmentation and melanoma. Here, we reveal a function for the metastasis-associated phosphatase of regenerating liver 3 (PRL3) in MSC regeneration. We show that PRL3 binds to the RNA helicase DDX21, thereby restricting productive transcription by RNAPII at master transcription factor (MITF)-regulated endolysosomal vesicle genes. In zebrafish, this mechanism controls premature melanoblast expansion and differentiation from MSCs. In melanoma patients, restricted transcription of this endolysosomal vesicle pathway is a hallmark of PRL3-high melanomas. Our work presents the conceptual advance that PRL3-mediated control of transcriptional elongation is a differentiation checkpoint mechanism for activated MSCs and has clinical relevance for the activity of PRL3 in regenerating tissue and cancer.
Sigma-Aldrich
AG 1478, A cell-permeable, reversible, ATP-competitive, highly potent and selective inhibitor of epidermal growth factor receptor kinase versus HER2-neu and platelet-derived growth factor receptor kinase.
Sigma-Aldrich
PRL-3 Inhibitor I, ≥98% (HPLC), solid
Sigma-Aldrich
4-Thiouridine, ≥98%