Merck
  • ATAD5 promotes replication restart by regulating RAD51 and PCNA in response to replication stress.
ATAD5 promotes replication restart by regulating RAD51 and PCNA in response to replication stress.
Nature communications (2019-12-18)
Su Hyung Park, Nalae Kang, Eunho Song, Minwoo Wie, Eun A Lee, Sunyoung Hwang, Deokjae Lee, Jae Sun Ra, In Bae Park, Jieun Park, Sukhyun Kang, Jun Hong Park, Sungchul Hohng, Kyoo-Young Lee, Kyungjae Myung
ABSTRACT
Maintaining stability of replication forks is important for genomic integrity. However, it is not clear how replisome proteins contribute to fork stability under replication stress. Here, we report that ATAD5, a PCNA unloader, plays multiple functions at stalled forks including promoting its restart. ATAD5 depletion increases genomic instability upon hydroxyurea treatment in cultured cells and mice. ATAD5 recruits RAD51 to stalled forks in an ATR kinase-dependent manner by hydroxyurea-enhanced protein-protein interactions and timely removes PCNA from stalled forks for RAD51 recruitment. Consistent with the role of RAD51 in fork regression, ATAD5 depletion inhibits slowdown of fork progression and native 5-bromo-2'-deoxyuridine signal induced by hydroxyurea. Single-molecule FRET showed that PCNA itself acts as a mechanical barrier to fork regression. Consequently, DNA breaks required for fork restart are reduced by ATAD5 depletion. Collectively, our results suggest an important role of ATAD5 in maintaining genome integrity during replication stress.
Sigma-Aldrich
Anti-Histone H3 Antibody, CT, pan, serum, Upstate®
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-Myc Tag Antibody, clone 4A6, clone 4A6, Upstate®, from mouse
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Triton X-100, laboratory grade