Merck
  • Dopamine D2 receptor modulates Wnt expression and control of cell proliferation.
Dopamine D2 receptor modulates Wnt expression and control of cell proliferation.
Scientific reports (2019-11-16)
Fei Han, Prasad Konkalmatt, Chaitanya Mokashi, Megha Kumar, Yanrong Zhang, Allen Ko, Zachary J Farino, Laureano D Asico, Gaosi Xu, John Gildea, Xiaoxu Zheng, Robin A Felder, Robin E C Lee, Pedro A Jose, Zachary Freyberg, Ines Armando
ABSTRACT
The Wnt/β-catenin pathway is one of the most conserved signaling pathways across species with essential roles in development, cell proliferation, and disease. Wnt signaling occurs at the protein level and via β-catenin-mediated transcription of target genes. However, little is known about the underlying mechanisms regulating the expression of the key Wnt ligand Wnt3a or the modulation of its activity. Here, we provide evidence that there is significant cross-talk between the dopamine D2 receptor (D2R) and Wnt/β-catenin signaling pathways. Our data suggest that D2R-dependent cross-talk modulates Wnt3a expression via an evolutionarily-conserved TCF/LEF site within the WNT3A promoter. Moreover, D2R signaling also modulates cell proliferation and modifies the pathology in a renal ischemia/reperfusion-injury disease model, via its effects on Wnt/β-catenin signaling. Together, our results suggest that D2R is a transcriptional modulator of Wnt/β-catenin signal transduction with broad implications for health and development of new therapeutics.
Sigma-Aldrich
(−)-Quinpirole hydrochloride, ≥98% (HPLC), solid
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Wnt3a
Sigma-Aldrich
MISSION® esiRNA, targeting human DRD2
Sigma-Aldrich
Anti-GAPDH Antibody, from chicken, purified by affinity chromatography
Sigma-Aldrich
Anti-Dopamine D2 Receptor Antibody, Chemicon®, from rabbit