Merck
  • Complete regeneration of large bone defects in rats with commercially available fibrin loaded with BMP-2.
Complete regeneration of large bone defects in rats with commercially available fibrin loaded with BMP-2.
European cells & materials (2019-09-19)
M Koolen, A Longoni, J van der Stok, O Van der Jagt, D Gawlitta, H Weinans
ABSTRACT
This study aimed at investigating in vitro and in vivo the efficiency of commercially available fibrin as a carrier for controlled and sustained bone morphogenetic protein-2 (BMP-2) release to induce bone formation and reduce the side effects of its use. In vitro release and activity of low-dose recombinant human BMP-2 (rhBMP-2) (37.5 µg/mL) embedded in commercially available fibrin were evaluated and, subsequently, critical-size femur defects in rats were grafted to study bone regeneration and vascularisation by micro-computed tomography (µCT) and histology. In vitro experiments showed a sustained BMP-2 release with a high BMP activity remaining after 28 d. In vivo, fibrin loaded with BMP-2 showed an extremely fast bone healing, with a large amount of new bone formation throughout the entire defect in the first 4 weeks and complete cortical repair and fusion after 8 weeks, with no ectopic bone formation. In contrast, the control fibrin group did not fuse after 12 weeks. Vascularisation was similar in both groups at 4 and 12 weeks after implantation. In conclusion, commercially available fibrin is a very efficient carrier for rhBMP-2 to graft critical-size cortical bone defects and might be a more optimal delivery vehicle for BMP-2-induced bone regeneration than currently available collagen sponges.
Sigma-Aldrich
Plasmin from human plasma, lyophilized powder, ≥2.0 units/mg protein
Sigma-Aldrich
Papaverine hydrochloride, powder
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg
Sigma-Aldrich
L-Ascorbic acid 2-phosphate sesquimagnesium salt hydrate, ≥95%
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
p-Nitrophenyl Phosphate Liquid Substrate System, liquid