Merck
  • Hepatocyte nuclear factor-1β regulates Wnt signaling through genome-wide competition with β-catenin/lymphoid enhancer binding factor.
Hepatocyte nuclear factor-1β regulates Wnt signaling through genome-wide competition with β-catenin/lymphoid enhancer binding factor.
Proceedings of the National Academy of Sciences of the United States of America (2019-11-13)
Siu Chiu Chan, Ying Zhang, Marco Pontoglio, Peter Igarashi
ABSTRACT
Hepatocyte nuclear factor-1β (HNF-1β) is a tissue-specific transcription factor that is essential for normal kidney development and renal tubular function. Mutations of HNF-1β produce cystic kidney disease, a phenotype associated with deregulation of canonical (β-catenin-dependent) Wnt signaling. Here, we show that ablation of HNF-1β in mIMCD3 renal epithelial cells produces hyperresponsiveness to Wnt ligands and increases expression of Wnt target genes, including Axin2, Ccdc80, and Rnf43 Levels of β-catenin and expression of Wnt target genes are also increased in HNF-1β mutant mouse kidneys. Genome-wide chromatin immunoprecipitation sequencing (ChIP-seq) in wild-type and mutant cells showed that ablation of HNF-1β increases by 6-fold the number of sites on chromatin that are occupied by β-catenin. Remarkably, 50% of the sites that are occupied by β-catenin in HNF-1β mutant cells colocalize with HNF-1β-occupied sites in wild-type cells, indicating widespread reciprocal binding. We found that the Wnt target genes Ccdc80 and Rnf43 contain a composite DNA element comprising a β-catenin/lymphoid enhancer binding factor (LEF) site overlapping with an HNF-1β half-site. HNF-1β and β-catenin/LEF compete for binding to this element, and thereby HNF-1β inhibits β-catenin-dependent transcription. Collectively, these studies reveal a mechanism whereby a transcription factor constrains canonical Wnt signaling through direct inhibition of β-catenin/LEF chromatin binding.
Sigma-Aldrich
Anti-RNF43 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-CCDC80 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution