Merck
  • Volatiles from the Mandibular Gland Reservoir Content of Colobopsis explodens Laciny and Zettel, 2018, Worker Ants (Hymenoptera: Formicidae).
Volatiles from the Mandibular Gland Reservoir Content of Colobopsis explodens Laciny and Zettel, 2018, Worker Ants (Hymenoptera: Formicidae).
Molecules (Basel, Switzerland) (2019-09-27)
Michaela Hoenigsberger, Alexey G Kopchinskiy, Christoph Bueschl, Alexandra Parich, Alice Laciny, Herbert Zettel, Kamariah A Salim, Linda Bl Lim, Irina S Druzhinina, Rainer Schuhmacher
ABSTRACT
Forty-five volatile organic compounds (VOCs) were identified or annotated in the mandibular gland reservoir content (MGRC) of the Southeast Asian ant Colobopsis explodens Laciny and Zettel, 2018 (Hymenoptera: Formicidae), using headspace solid-phase microextraction (HS-SPME) coupled to gas chromatography mass spectrometry (GC-MS) and liquid extraction combined with GC-MS. In extension of previous reports on VOCs of C. explodens, members of different compound classes, such as alkanes, aliphatic and aromatic carboxylic acids, and phenolics, were detected. The ketone 2-heptanone and the biochemically related phenolics benzene-1,3,5-triol (phloroglucinol, PG), 1-(2,4,6-trihydroxyphenyl)ethanone (monoacetylphloroglucinol, MAPG), 5,7-dihydroxy-2-methylchromen-4-one (noreugenin), and 1-(3-Acetyl-2,4,6-trihydroxyphenyl)ethanone (2,4-diacetylphloroglucinol, DAPG) dominated the GC-MS chromatograms. The identities of the main phenolics MAPG and noreugenin were further verified by liquid chromatography-high resolution-tandem mass spectrometry (LC-HRMS/MS). A comparative study of MGRC samples originating from three distinct field expeditions revealed differences in the VOC profiles, but the presence and relative abundances of the dominating constituents were largely consistent in all samples. Our study considerably extends the knowledge about the number and type of VOCs occurring in the MGRC of C. explodens. Based on the type of the detected compounds, we propose that the likely irritant and antibiotic phenolic constituents play a role in defense against arthropod opponents or in protection against microbial pathogens.
Sigma-Aldrich
2-Heptanone, 99%
Sigma-Aldrich
Dehydroacetic acid, ≥98.0% (T)
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
1-Heptadecene, 98%
Sigma-Aldrich
1,2,4-Trimethylbenzene, 98%
Sigma-Aldrich
Orcinol, 97%
Sigma-Aldrich
1-Undecene, 97%
Sigma-Aldrich
1-Pentadecene, 98%
Sigma-Aldrich
Nonanoic acid, ≥97%
Sigma-Aldrich
4-Hydroxy-6-methyl-2-pyrone, 98%
Sigma-Aldrich
Mesitylene, reagent grade, 97%