Merck
  • Tetramethylpyrazine prevents diabetes by activating PI3K/Akt/GLUT-4 signalling in animal model of type-2 diabetes.
Tetramethylpyrazine prevents diabetes by activating PI3K/Akt/GLUT-4 signalling in animal model of type-2 diabetes.
Life sciences (2019-09-08)
Uddipak Rai, Ramoji Kosuru, Swati Prakash, Surya Pratap Singh, Hareram Birla, Vinod Tiwari, Sanjay Singh
ABSTRACT
The present experiment was conceptualised to explore the therapeutic response of tetramethylpyrazine (TMP), a major active constituent of Ligusticum chuanxiong, a Chinese traditional medicinal plant, in high-fat diet (HFD)-streptozotocin (STZ)-induced diabetes in rats and to identify the possible mechanism of action. Dose-reliant effect of oral treatment of TMP (100, 150 and 200 mg/kg/day) for 28 days was evaluated by calculating the alteration in body weight, level of fasting blood glucose (FBG), plasma insulin, homeostasis model assessment (HOMA), serum lipids, oral glucose & intraperitoneal insulin tolerance and glycosylated haemoglobin in HFD-STZ-induced type-2 diabetic (T2D) rats and underlying molecular mechanisms of TMP was also studied. TMP treatment prominently reduced the level of FBG, glycosylated haemoglobin and revived body weight gain and level of serum insulin dose-dependently in diabetic rats. TMP treatment considerably improved insulin resistance, as observed in oral glucose tolerance and insulin tolerance tests. Moreover, dose-dependent reduction in the level of pro-inflammatory cytokines, C-reactive protein (CRP) and interleukin-6 (IL-6) was observed and their level was found to be significantly reduced in highest dose TMP (200 mg/kg) treated diabetic rats, pointing towards TMP mediated recovery of insulin signalling and a decrease in insulin resistance. The expressions of p-PI3K-p85/p-Akt/GLUT-4 were also significantly up-regulated by TMP (200 mg/kg), suggesting the connection of the PI3K/Akt signal pathway in the anti-hyperglycemic action of TMP. These findings suggest that TMP may be used as a potential agent for type-2 diabetes treatment.
Sigma-Aldrich
Rat IL-6 ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
Rat CRP/C-Reactive Protein ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Rat Ins1 / Insulin ELISA Kit
Sigma-Aldrich
2,3,5,6-Tetramethylpyrazine, natural, ≥98%, FG
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder