Merck
  • Adaptive disinhibitory gating by VIP interneurons permits associative learning.
Adaptive disinhibitory gating by VIP interneurons permits associative learning.
Nature neuroscience (2019-10-23)
Sabine Krabbe, Enrica Paradiso, Simon d'Aquin, Yael Bitterman, Julien Courtin, Chun Xu, Keisuke Yonehara, Milica Markovic, Christian Müller, Tobias Eichlisberger, Jan Gründemann, Francesco Ferraguti, Andreas Lüthi
ABSTRACT
Learning drives behavioral adaptations necessary for survival. While plasticity of excitatory projection neurons during associative learning has been extensively studied, little is known about the contributions of local interneurons. Using fear conditioning as a model for associative learning, we found that behaviorally relevant, salient stimuli cause learning by tapping into a local microcircuit consisting of precisely connected subtypes of inhibitory interneurons. By employing deep-brain calcium imaging and optogenetics, we demonstrate that vasoactive intestinal peptide (VIP)-expressing interneurons in the basolateral amygdala are activated by aversive events and provide a mandatory disinhibitory signal for associative learning. Notably, VIP interneuron responses during learning are strongly modulated by expectations. Our findings indicate that VIP interneurons are a central component of a dynamic circuit motif that mediates adaptive disinhibitory gating to specifically learn about unexpected, salient events, thereby ensuring appropriate behavioral adaptations.
Sigma-Aldrich
Anti-Red Fluorescent Protein Antibody, Chemicon®, from chicken
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat
Sigma-Aldrich
Anti-Somatostatin Antibody, clone YC7, culture supernatant, clone YC7, Chemicon®
Sigma-Aldrich
Triton X-100, laboratory grade