Merck
  • DNA Aptamers Targeting BACE1 Reduce Amyloid Levels and Rescue Neuronal Deficiency in Cultured Cells.
DNA Aptamers Targeting BACE1 Reduce Amyloid Levels and Rescue Neuronal Deficiency in Cultured Cells.
Molecular therapy. Nucleic acids (2019-04-09)
Jun Xiang, Wen Zhang, Xiao-Fang Cai, Min Cai, Zhong-Hai Yu, Feng Yang, Wen Zhu, Xiang-Ting Li, Ting Wu, Jing-Si Zhang, Ding-Fang Cai
ABSTRACT
β-amyloid (Aβ) plays an essential role in the pathogenesis of Alzheimer's disease (AD). Beta-site amyloid precursor protein cleaving enzyme 1 (BACE1) is indispensable for Aβ production, and knockout of BACE1 has no overt phenotypes in mouse. Thus, fine modulation of BACE1 may be a safe and effective treatment for AD patients. However, the large active site of BACE1 makes it challenging to target BACE1 with classical small-molecule inhibitors. DNA aptamer can have high affinity and specificity against diverse targets, and it provides an alternative strategy to target BACE1. In this study, we used a novel cell-systematic evolution of ligands by exponential enrichment (SELEX) strategy to select specific DNA aptamers optimized to target BACE1 under physiological status. After 17 rounds of selection, we identified two DNA aptamers against BACE1: BI1 and BI2. The identified aptamers interacted with BACE1 in pull-down assay, inhibited BACE1 activity in in vitro fluorescence resonance energy transfer (FRET) assay and HEK293-APP stable cell line, reduced Aβ in the culture medium of HEK293-amyloid protein precursor (APP) stable cell line and APP-PS1 primary cultured neurons, and rescued Aβ-induced neuronal deficiency in APP-PS1 primary cultured neurons. In contrast, the identified aptamers had no effect on α- or γ-secretase. In addition, cholesteryl tetraetylene glycol (TEG) modification further improved the potency of the identified aptamers. Our study suggests that it is feasible and effective to target BACE1 with DNA aptamers, and the therapeutic potential of the identified aptamers deserves further investigation.
Sigma-Aldrich
Anti-BACE Antibody, CT, clone 61-3E7, clone 61-3E7, Chemicon®, from mouse
Sigma-Aldrich
Anti-GluR1 Antibody, clone C3T, rabbit monoclonal, culture supernatant, clone C3T, Upstate®
Sigma-Aldrich
γ-Secretase Inhibitor IX, Gamma-Secretase Inhibitor IX - CAS 208255-80-5, is a cell-permeable inhibitor of γ-secretase (Aβtotal IC₅₀ = 115 nM, Aβ42 IC₅₀ = 200 nM).
Sigma-Aldrich
Anti-Synaptophysin Antibody, clone SY38, clone SY38, Chemicon®, from mouse
Sigma-Aldrich
β-Secretase Inhibitor IV, β-Secretase Inhibitor IV, CAS 797035-11-1, is a cell-permeable inhibitor that binds to BACE-1 active site and blocks its proteolytic activity (IC₅₀ = 15 nM for human BACE-1).
Sigma-Aldrich
Anti-Synapsin I antibody produced in rabbit, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Anti-PSD95 Antibody, clone K28/43, clone K28/43, from mouse