Merck
  • TOM40 Targets Atg2 to Mitochondria-Associated ER Membranes for Phagophore Expansion.
TOM40 Targets Atg2 to Mitochondria-Associated ER Membranes for Phagophore Expansion.
Cell reports (2019-08-15)
Zhenyuan Tang, Yoshinori Takahashi, Haiyan He, Tatsuya Hattori, Chong Chen, Xinwen Liang, Han Chen, Megan M Young, Hong-Gang Wang
ABSTRACT
During autophagy, phagophores grow into double-membrane vesicles called autophagosomes, but the underlying mechanism remains unclear. Here, we show a critical role of Atg2A in phagophore expansion. Atg2A translocates to the phagophore at the mitochondria-associated ER membrane (MAM) through a C-terminal 45-amino acid domain that we have termed the MAM localization domain (MLD). Proteomic analysis identifies the outer mitochondrial membrane protein TOM40 as a MLD-interacting partner. The Atg2A-TOM40 interaction is responsible for MAM localization of Atg2A and requires the TOM receptor protein TOM70. In addition, Atg2A interacts with Atg9A by a region within its N terminus. Inhibition of either Atg2A-TOM40 or Atg2A-Atg9A interactions impairs phagophore expansion and accumulates Atg9A-vesicles in the vicinity of autophagic structures. Collectively, we propose a model that the TOM70-TOM40 complex recruits Atg2A to the MAM for vesicular and/or non-vesicular lipid transport into the expanding phagophore to grow the size of autophagosomes for efficient autophagic flux.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid