Merck
IRF4 haploinsufficiency in a family with Whipple's disease.
eLife (2018-03-15)
Antoine Guérin, Gaspard Kerner, Nico Marr, Janet G Markle, Florence Fenollar, Natalie Wong, Sabri Boughorbel, Danielle T Avery, Cindy S Ma, Salim Bougarn, Matthieu Bouaziz, Vivien Béziat, Erika Della Mina, Carmen Oleaga-Quintas, Tomi Lazarov, Lisa Worley, Tina Nguyen, Etienne Patin, Caroline Deswarte, Rubén Martinez-Barricarte, Soraya Boucherit, Xavier Ayral, Sophie Edouard, Stéphanie Boisson-Dupuis, Vimel Rattina, Benedetta Bigio, Guillaume Vogt, Frédéric Geissmann, Lluis Quintana-Murci, Damien Chaussabel, Stuart G Tangye, Didier Raoult, Laurent Abel, Jacinta Bustamante, Jean-Laurent Casanova
ABSTRACT
Most humans are exposed to Tropheryma whipplei (Tw). Whipple's disease (WD) strikes only a small minority of individuals infected with Tw (<0.01%), whereas asymptomatic chronic carriage is more common (<25%). We studied a multiplex kindred, containing four WD patients and five healthy Tw chronic carriers. We hypothesized that WD displays autosomal dominant (AD) inheritance, with age-dependent incomplete penetrance. We identified a single very rare non-synonymous mutation in the four patients: the private R98W variant of IRF4, a transcription factor involved in immunity. The five Tw carriers were younger, and also heterozygous for R98W. We found that R98W was loss-of-function, modified the transcriptome of heterozygous leukocytes following Tw stimulation, and was not dominant-negative. We also found that only six of the other 153 known non-synonymous IRF4 variants were loss-of-function. Finally, we found that IRF4 had evolved under purifying selection. AD IRF4 deficiency can underlie WD by haploinsufficiency, with age-dependent incomplete penetrance.
Product Description
Roche
cOmplete Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Leupeptin, microbial, ≥90% (HPLC)
Sigma-Aldrich
Pepstatin A, microbial, ≥75% (HPLC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Hydrochloric acid, BioReagent, 36.5-38.0%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Triton X-100, laboratory grade