
Lansoprazole Related Compound B

Synonym(s):
2-[[[3-Methyl-4-(2,2,2-trifluoroethoxy)-pyridin-2-yl]methyl]sulfanyl]-1H-benzimidazole, Lansoprazole sulfide
Empirical Formula (Hill Notation):
C16H14F3N3OS
Molecular Weight:
353.36
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2