
Intermediates for Further Manufacturing Use (FFMU) - RHCE (Rh) Specificities

Products
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Quality Segment
Package Size
Technique
Showing 1-17 of 17
Compare
Product Number
Description
Pricing