
Intermediates For Further Manufacturing Use (FFMU) - Anti Human Globulin

Products
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Quality Segment
Package Size
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing