Skip to Content
Merck
Imidazolium bromide ≥98%

Imidazolium bromide

Imidazolium bromide ≥98%
Synonym(s):
Imidazole hydrobromide
Empirical Formula (Hill Notation):
C3H4N2 · HBr
CAS Number:
Molecular Weight:
148.99

Products

Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing