
Daunorubicin for system suitability

Synonym(s):
Daunorubicin hydrochloride, Daunomycin hydrochloride
Empirical Formula (Hill Notation):
C27H29NO10 · HCl
CAS Number:
Molecular Weight:
563.98
Products
Formula Weight
Manufacturer
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing