Skip to Content
Merck
cis-Dichlorobis(2,2′-bipyridine)ruthenium(II) hydrate 97%

cis-Dichlorobis(2,2′-bipyridine)ruthenium(II) hydrate

cis-Dichlorobis(2,2′-bipyridine)ruthenium(II) hydrate 97%
Synonym(s):
cis-Bis(2,2′-bipyridine)dichlororuthenium(II) hydrate
Empirical Formula (Hill Notation):
C20H16Cl2N4Ru · xH2O
CAS Number:
Molecular Weight:
484.34 (anhydrous basis)

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing