Skip to Content
Merck
4,5-Diamino-6-hydroxypyrimidine hemisulfate salt

4,5-Diamino-6-hydroxypyrimidine hemisulfate salt

4,5-Diamino-6-hydroxypyrimidine hemisulfate salt
Synonym(s):
5,6-Diamino-4-pyrimidinone hemisulfate salt, 5,6-Diamino-4-pyrimidinol hemisulfate salt
Empirical Formula (Hill Notation):
C8H12N8O2 · H2SO4
CAS Number:
Molecular Weight:
350.31

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing