biological source
mouse
conjugate
Atto 390 conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
2G4, monoclonal
form
buffered aqueous solution
mol wt
85kDa
species reactivity
rat, mouse
concentration
1.0 mg/mL
technique(s)
immunohistochemistry: 1:150
western blot: 1:1000
isotype
IgG2b
NCBI accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
rat ... Scnn1a(25122)
Immunogen
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service