Skip to Content
Merck
Sign Into View Organizational & Contract Pricing

About This Item

Empirical Formula (Hill Notation):
FeO4S · 7H2O
CAS Number:
Molecular Weight:
278.01
MDL number:
UNSPSC Code:
12352302
Grade:
for analytical purposes
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

for analytical purposes

SMILES string

[Fe+2].[S](=O)(=O)([O-])[O-].O.O.O.O.O.O.O

InChI

1S/Fe.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

InChI key

SURQXAFEQWPFPV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service