Skip to Content
Merck
Cerium(IV) oxide-samarium doped <0.5 μm particle size, powder, contains 20 mol % samarium as dopant

Samarium-doped ceria

Cerium(IV) oxide-samarium doped <0.5 μm particle size, powder, contains 20 mol % samarium as dopant
Synonym(s):
SDC, Samarium-doped ceria
Empirical Formula (Hill Notation):
CeO2
Molecular Weight:
172.11

Products

Sustainability
Feature
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Particle Size (µm)
Showing 1-1 of 1