
Non Animal Origin Squalene

Synonym(s):
Squalene
Empirical Formula (Hill Notation):
C30H50
CAS Number:
Molecular Weight:
410.72
Products
Product Category
Sustainability
Formula Weight
Physical Form
Product Line
Agency
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing